Order Entry
Singapore
ContactUsLinkComponent
1-Bromo-2-(bromomethyl)naphthalene ≥98%
1-Bromo-2-(bromomethyl)naphthalene ≥98%
Catalog # ALFAH50240.03
CAS Number:  
1-Bromo-2-(bromomethyl)naphthalene ≥98%
Catalog # ALFAH50240.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:37763-43-2
 :
Formula:C₁₁H₈Br₂
MDL Number:MFCD00046369
 :299.99 g/mol
Melting Pt:108 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05