Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Phenyl-1H-pyrazole-4-carboxylic acid
1-Phenyl-1H-pyrazole-4-carboxylic acid ≥99%
1-Phenyl-1H-pyrazole-4-carboxylic acid ≥99%
Irritant
1-Phenyl-1H-pyrazole-4-carboxylic acid ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
1-Phenyl-1H-pyrazole-4-carboxylic acid ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
1134-50-5
:
Formula:
C₁₀H₈N₂O₂
MDL Number:
MFCD00514518
:
188.19 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH50305
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation