Order Entry
Singapore
ContactUsLinkComponent
4-Anilino-1-methylpiperidine ≥98%
Toxic
Irritant
4-Anilino-1-methylpiperidine ≥98%
Catalog # ALFAH27276.14
CAS Number:  
Toxic
Irritant
4-Anilino-1-methylpiperidine ≥98%
Catalog # ALFAH27276.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

CAS Number:22261-94-5
 :
Formula:C₁₂H₁₈N₂
MDL Number:MFCD06408133
 :190.28 g/mol
Melting Pt:82…86 °C
EINECS:244-882-7
 :GHS06
 :GHS07
Storage Temperature:Ambient