Order Entry
Singapore
ContactUsLinkComponent
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')holmium(III)
Irritant
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')holmium(III)
Catalog # ALFA33531.06
CAS Number:  
Irritant
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')holmium(III)
Catalog # ALFA33531.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :714.732 g/mol
Melting Pt:179…183 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:15522-73-3
Formula:C₃₃H₅₇HoO₆
MDL Number:MFCD00070457