Order Entry
Singapore
ContactUsLinkComponent
Blood Bank Refrigerators
Blood Bank Refrigerators
Supplier:  REVCO TECHNOLOGIES
Certificates
undefined
Blood Bank Refrigerators
Supplier:  REVCO TECHNOLOGIES
Specifications
  • Catalog No:
  • REVCREB404V
  • REVCREB5004V
  • REVCREB1204V
  • REVCREB2304V
  • Model:
  • REB-404V
  • REB-5004V
  • REB-1204V
  • REB-2304V
  • Cat. No.:
  • REVCREB404V
  • REVCREB5004V
  • REVCREB1204V
  • REVCREB2304V
  • Air circulation:
  • Ventilated
  • Ventilated
  • Ventilated
  • Ventilated
  • Display:
  • Digital
  • Digital
  • Digital
  • Digital
  • Defrost:
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Alarm:
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Access ports:
  • 1
  • 1
  • 1
  • 1
  • Shelves/drawers:
  • 2 shelves
  • 12 shelves
  • 5 shelves
  • 6 shelves
  • Door type:
  • 1 solid swing door
  • 2 glass swing doors
  • 1 glass swing door
  • 1 glass swing door
  • Door lock:
  • Yes
  • Yes
  • Yes
  • Yes
  • Material:
  • Steel
  • Steel
  • Steel
  • Steel
  • Weight:
  • 96 kg
  • 359 kg
  • 145 kg
  • 213 kg
Specifications