Tubing, silicone, EXACANAL
Supplier: Saint Gobain Life Sciences
Certificates
About this item
Silicone COVOSIL 9161, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
- Very flexible
Temperature resistance: –60 to +200 °C
Hardness (Shore durometer A): 60
Density: 1,14
Certifications: Complies with FDA (21 CFR § 177.2600) standards