Order Entry
Singapore
Orders LinkContactUsLinkComponent
SHEL-LAB-Microbiological-Incubators955-590.jpg
SHEL LAB Microbiological Incubators
SHEL LAB Microbiological Incubators | Avantor
Microbiological incubator series has been updated for optimum performance. Ranging in sizes from 57 to 331 liters (2 to 11.7 cu.ft.), the SMI series is guaranteed to fit any workspace. Ideal for clinical, analytical, and general laboratory applications.
  • USB data logging capabilities
  • Independent over temperature set point and operational control override for additional safety
  • Side-mounted access port, 38 mm diameter (1.5")
  • Stainless steel interior construction for long life operation and easy cleaning
  • Condensed connection panel includes; 4 to 20 mA analog output, dry contact port, and USB A port
  • Available in 100 to 120 V or 220 to 240 V configurations - 24 month limited warranty
  • Perfect solution for general cell culture and constant temperature applications
Heated external doors prevent cold spots and maintain thermal uniformity. Unique air-jacketed design provides excellent temperature uniformity of ±0.5 °C at 37 °C. Interior chamber outlet (1 amp). Sealed glass inner door allows for viewing without disturbing the critical growth atmosphere.
Capacitive touch screen of 7" usable with laboratory gloves. With on-screen graphing and real time data, the new SMI controller provides the highest levels of sample security and traceability. Touch screen controller features: Storage up to 5 years of log data, password locking capabilities, data logged once per minute, export in .CSV file, smart alarm (accounts for door openings and temperature set point changes before alarm), temperature deviation alarm.
Certifications: Safety certified CAN/CSA, TUV, UL, EN, IEC 61010 and compliant with CE.
SMI2-Incubator400-495.jpg
SMI2 Microbiological Incubator - 56.3 Liter
The SHEL LAB Microbiological Incubator Series is designed for optimum performance.
SMI6-Incubator400-495.jpg
SMI6 Microbiological Incubator - 166 Liter
The SHEL LAB Microbiological Incubator Series is designed for optimum performance.
SMI7-Incubator400-495.jpg
SMI7 Microbiological Incubator - 184 Liter
The SHEL LAB Microbiological Incubator Series is designed for optimum performance.
SMI11-Incubator400-495.jpg
SMI11 Microbiological Incubator - 309 Liter
The SHEL LAB Microbiological Incubator Series is designed for optimum performance.