Orders LinkContactUsLinkComponent
Bravo Disposable Pipette Tips
Bravo Disposable Pipette Tips
384 in rack, wide bore sterile filtered, conductive, 70 µl, Compatible with Bravo 96ST or 384ST head, 50/Case
Katalog # AGLS19134-022
Leverantör:   AGILENT
Bravo Disposable Pipette Tips
Katalog # AGLS19134-022
Leverantör:  AGILENT
Bravo Disposable Pipette Tips
Bravo Disposable Pipette Tips
384 in rack, wide bore sterile filtered, conductive, 70 µl, Compatible with Bravo 96ST or 384ST head, 50/Case
Price
50 741,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 19.200 Items
Product Details & Documents
Certified on Bravo and AssayMAP for accuracy and precision.
  • Individual lot certified to meet thorough QC standards
  • Certified free of detectable DNase and RNase activity
  • Certified free of endotoxin (pyrogen) contamination
  • Certified free of adenosine triphosphate (ATP)
  • Certified free of human DNA;Certified free of protease activity
  • Certified free of heavy metals
  • Certified as sterile with a sterility assurance level (SAL) of 10⁻⁶
  • Innovative processing and packaging reduce static buildup and improve deck space utilisation
Agilent offers disposable pipette tips in both 96- and 384-well formats specially designed, tested, and optimized for the Bravo Automated Liquid Handling Platform and the AssayMAP Bravo heads. All Agilent pipette tips undergo complete QC and functional performance testing for each lot.
The innovative processing and packaging of Bravo robotic tips helps address common concerns about lab disposable pipette tips used in automation, such as static buildup and deck space utilisation. Additionally, the Agilent Bravo disposable pipette tips are the only pipette tips that are certified for use on Agilent Bravo Liquid Handling instrumentation, with the assurance that every tip meets our exacting accuracy and precision requirements.
Förpackning: Packed as case of 50, except: AGLS21740-202, AGLS21740-212, AGLS21750-202 and AGLS21750-212 which are a case of 90.
Specifikationer
  • Förp.:
    50/Case
  • Volym:
    70 µl
  • Seq:
    21
  • Tip style:
    384 in rack, wide bore sterile filtered, conductive
  • För användning med:
    Compatible with Bravo 96ST or 384ST head