Täckglas
8×8 mm, #2, Glas
Katalog # MARI0103990091
Leverantör: MARIENFELD
Gå till:
Product Details & Documents
Made of chemically resistant borosilicate D 263™ M glass, Hydrolytic Class I
- No bubbles or flaws
- Suitable for in vitro diagnostic (IVD) applications
Thickness: 0,13 - 0,16 mm
: Supplied 100 in hinged boxes or in boxes with separate lid.
Certificates
Sök efter LOT-nummerSpecifikationer
- Nr:#2
- L×B:8×8 mm
- Material:Glas
- Seq:01