Order Entry
Sweden
Orders LinkContactUsLinkComponent
EJ SE

EJ SE

Blood sampling, made in china, with Max.shelf life 60 months, 19, 30 mm, 1,10 mm, Gräddfärgad
Katalog # BRAU4665317B
Leverantör:   B. Braun
EJ SE
Katalog # BRAU4665317B
Leverantör:  B. Braun
EJ SE
EJ SE

Blood sampling, made in china, with Max.shelf life 60 months, 19, 30 mm, 1,10 mm, Gräddfärgad

Price
120,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 Items
Product Details & Documents

Stainless, chromium-nickel steel with transparent PP Luer lock plastic hub. These sterile hypodermic needles Sterican® have the following features: Suitable for single-use hypodermic needles for intra-muscular, subcutaneous, intravenous, intra-arterial and special injections. Thin walled needles.

  • Extremely smooth surface with light silicone coating
  • 3-facet tips, long or short bevel
  • For minimal pain upon puncture

 : ISO 6009

Specifikationer
  • Färgkod:
    Gräddfärgad
  • Ø:
    1,10 mm
  • Mätare:
    19
  • Beskrivning:
    Blood sampling, made in china, with Max.shelf life 60 months
  • Längd:
    30 mm
  • Seq:
    02