Order Entry
Sweden
Orders LinkContactUsLinkComponent
EJ SE

EJ SE

Blood sampling, made in china, with Max.shelf life 60 months, 19, 40 mm, 1,10 mm, Gräddfärgad
Catalog # BRAU4657799B
Supplier:   B. Braun
EJ SE
Catalog # BRAU4657799B
Supplier:  B. Braun
EJ SE
EJ SE

Blood sampling, made in china, with Max.shelf life 60 months, 19, 40 mm, 1,10 mm, Gräddfärgad

Price
120,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents

Stainless, chromium-nickel steel with transparent PP Luer lock plastic hub. These sterile hypodermic needles Sterican® have the following features: Suitable for single-use hypodermic needles for intra-muscular, subcutaneous, intravenous, intra-arterial and special injections. Thin walled needles.

  • Extremely smooth surface with light silicone coating
  • 3-facet tips, long or short bevel
  • For minimal pain upon puncture

Certifications: ISO 6009

Specifications
  • Färgkod:
    Gräddfärgad
  • Ø:
    1,10 mm
  • Mätare:
    19
  • Beskrivning:
    Blood sampling, made in china, with Max.shelf life 60 months
  • Längd:
    40 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.