Order Entry
Sweden
Orders LinkContactUsLinkComponent
EJ SE

EJ SE

Blood sampling, phlebotomy, with short bevel, made in china, with Max.shelf life 60 months, 18, 40 mm, 1,20 mm, Rosa
Katalog # BRAU4665120B
Leverantör:   B. Braun
EJ SE
Katalog # BRAU4665120B
Leverantör:  B. Braun
EJ SE
EJ SE

Blood sampling, phlebotomy, with short bevel, made in china, with Max.shelf life 60 months, 18, 40 mm, 1,20 mm, Rosa

Price
120,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 Items
Product Details & Documents

Stainless, chromium-nickel steel with transparent PP Luer lock plastic hub. These sterile hypodermic needles Sterican® have the following features: Suitable for single-use hypodermic needles for intra-muscular, subcutaneous, intravenous, intra-arterial and special injections. Thin walled needles.

  • Extremely smooth surface with light silicone coating
  • 3-facet tips, long or short bevel
  • For minimal pain upon puncture

Certifications: ISO 6009

Specifikationer
  • Färgkod:
    Rosa
  • Ø:
    1,20 mm
  • Mätare:
    18
  • Beskrivning:
    Blood sampling, phlebotomy, with short bevel, made in china, with Max.shelf life 60 months
  • Längd:
    40 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.