Rectengular glass capillaries made of Borosilicate glass, one side closed Length 100 ± 1 mm, Wallthickness about 120 microns Outer hight 4,2 ± 0,2 mm outer width 1,25 ± 0,05 mm 1 * 25 Items
Rectengular glass capillaries made of Borosilicate glass, one side closed Length 100 ± 1 mm, Wallthickness about 120 microns Outer hight 4,2 ± 0,2 mm outer width 1,25 ± 0,05 mm 1 * 25 Items