Orders LinkContactUsLinkComponent
QuickConnect™ Mop Handles
QuickConnect™ Mop Handles
QuickConnect™ carbon fiber telescopic handle, 160 - 290 cm (63 - 114")
Katalog # CONTCFH114
Leverantör:   CONTEC
QuickConnect™ Mop Handles
Katalog # CONTCFH114
Leverantör:  CONTEC
QuickConnect™ Mop Handles
QuickConnect™ Mop Handles
QuickConnect™ carbon fiber telescopic handle, 160 - 290 cm (63 - 114")
Price
3 025,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, as well as anodised aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths.
Specifikationer
  • Beskrivning:
    QuickConnect™ carbon fiber telescopic handle
  • Längd:
    160 - 290 cm (63 - 114")
  • Seq:
    03