Cerex® THC SPE Columns
Cerex Atlas® column THC, THC, 80 Å, 17 µm, 1 ml, 10 mg
Catalog # TCAT682-0101R
Supplier: Tecan
Jump to:
Product Details & Documents
Polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation. Specially formulated for THC/THCA testing.
Specifications
- Partikelstorlek:17 µm
- Porstorlek:80 Å
- Kapacitet:1 ml
- Beskrivning:Cerex Atlas® column THC
- Seq:01
- Bäddvikt:10 mg
- Phase:THC