Resolvex® Column Temperature Controller
Column heater 250 CERA, 1,5×1,5×17"
Katalog # TCAT282-0252
Leverantör: Tecan
Gå till:
Product Details & Documents
Column temperature controllers for heating and cooling.
- Reliable PID microprocessor controlled thermal circuit stabilizes the operating environment
- Easily-seated thermal blanket encloses the column in horizontal, vertical or slant orientation
- Heater inserts are modular for easy removal and arrangement
Based on a unique, space-saving concept for efficient temperature control of liquid chromatography columns. Column enclosure accepts ³/₈" ext. Ø HPLC columns of any length up to 30 centimeters (15 centimeters in Column Heater 150).
Specifikationer
- Beskrivning:Column heater 250 CERA, 1,5×1,5×17"
- Seq:02