SERAspher HPLC Columns, BioRP
Analytical, Stainless steel, bioinert coated, 100 Å, 5 µm, 7,8 mm, 300 mm
Katalog # ISERA1P2-A25L0B
Leverantör: ISERA
Gå till:
Product Details & Documents
Ideal for separation and purification of biomolecules.
- Polymeric particle with no silanol interactions and high chemical and thermal stability
- High performance in peptide and biochromatography
Used in bioscience, pharmaceutical industries, clinical research, environmental, food and agriculture with reproducable results and long lifetime.
Certifications: Individual test certificate is enclosed with each column.
Leveransinformation: Columns are delivered in solvent which is suitable for reversed phase chromatography.
Specifikationer
- Kolumnformat:Analytical
- Ø inr.:7,8 mm
- Partikelstorlek:5 µm
- Porstorlek:100 Å
- Längd:300 mm
- Seq:27
- Column hardware:Stainless steel, bioinert coated
- USP code:L21
- Max. tryck:200 bar
- Cat. No.:ISERA1P2-A25L0B
- pH område:1 - 14
- Förvaring:RT
- Applikation:Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
- Phase name:SERAspher
- Shipping conditions:Shipped under testing eluent.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



