SERAspher HPLC Columns, BioRP
Analytical, Stainless steel, bioinert coated, 100 Å, 5 µm, 4,6 mm, 250 mm
Katalog # ISERA1P1-A25Y0B
Leverantör: ISERA
Gå till:
Product Details & Documents
Ideal for separation and purification of biomolecules.
- Polymeric particle with no silanol interactions and high chemical and thermal stability
- High performance in peptide and biochromatography
Used in bioscience, pharmaceutical industries, clinical research, environmental, food and agriculture with reproducable results and long lifetime.
: Individual test certificate is enclosed with each column.
: Columns are delivered in solvent which is suitable for reversed phase chromatography.
Specifikationer
- Kolumnformat:Analytical
- Ø inr.:4,6 mm
- Partikelstorlek:5 µm
- Porstorlek:100 Å
- Längd:250 mm
- Seq:24
- Column hardware:Stainless steel, bioinert coated
- Cat. No.:ISERA1P1-A25Y0B
- USP code:L21
- pH område:1 - 14
- Max. tryck:200 bar
- Förvaring:RT
- Applikation:Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
- Phase name:SERAspher
- Shipping conditions:Shipped under testing eluent.



