Self-laminating Polyester Labels for M710
Labels, rectangular, B-461, M7-119-461, Polyester, Permanent akryl, White/Transparent, M710, 15,24W×48,26H mm, 500/box
Katalog # BRDY311979
Leverantör: Brady
Gå till:
Product Details & Documents
Self-laminating polyester labels for M710 printer.
- Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
- Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.
Specifikationer
- Printer model:M710
- Typ:B-461
- Färg:White/Transparent
- Beskrivning:Labels, rectangular
- Mått:15,24W×48,26H mm
- Material:Polyester
- Seq:04
- Modell:M7-119-461
- Förp.:500/box
- Självhäftande:Permanent akryl