Self-laminating Polyester Labels for M710
Labels, rectangular and round, B-101, M7-101-461, Polyester, Permanent akryl, White/Transparent, M710, 40,00W×15,00H×9,53Ø m, 250 sets/box
Catalog # BRDY311974
Supplier: Brady
Jump to:
Product Details & Documents
Self-laminating polyester labels for M710 printer.
- Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
- Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.
Specifications
- Printer model:M710
- Typ:B-101
- Färg:White/Transparent
- Beskrivning:Labels, rectangular and round
- Mått:40,00W×15,00H×9,53Ø m
- Material:Polyester
- Seq:09
- Modell:M7-101-461
- Förp.:250 sets/box
- Självhäftande:Permanent akryl