Orders LinkContactUsLinkComponent
Bioanalyzer RNA Analysis
Bioanalyzer RNA Analysis
Agilent RNA 6000 nano kit, For the analysis and quantitation of total and mRNA samples of 25 to 500 ng/µl in concentration, the kit includes 25 microfluidic chips, reagents and consumables, sufficient for 300 samples
Katalog # AGLS5067-1511
Leverantör:   AGILENT
Bioanalyzer RNA Analysis
Katalog # AGLS5067-1511
Leverantör:  AGILENT
Bioanalyzer RNA Analysis
Agilent RNA 6000 nano kit, For the analysis and quantitation of total and mRNA samples of 25 to 500 ng/µl in concentration, the kit includes 25 microfluidic chips, reagents and consumables, sufficient for 300 samples
Price
11 046,71 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Reliable and reproducible RNA sample quality assessment.
  • Maximum flexibility is enabled with options to check total RNA, mRNA, or cRNA quality
  • Minimal sample consumption is required with only 5 ng of total RNA needed for an effective analysis
  • Easily determine the RNA integrity number (RIN) using in-built software that automatically assigns an integrity value to a total RNA sample
  • This tool was designed to provide scientists an objective measurement of degradation in RNA samples
  • RNase free consumables avoid sample degradation during analysis
  • Accurate results are achieved rapidly, with 12 samples analysed in approximately 30 minutes
The Bioanalyzer RNA 6000 Nano assay provides reliable and reproducible characterization of total RNA and mRNA from multiple sample types, from as low as 5 ng of total RNA. The software will provide an objective measurement of RNA quality with RIN (RNA Integrity Number). Validated RIN values are available for eukaryotic, prokaryotic, and plant samples.
The Bioanalyzer RNA kits allow dependable and precise integrity checks and sample quantitation prior to any RNA-dependent application. This ensures the data from downstream research processes, such as gene expression microarray, qPCR, and transcriptome sequencing, are valid and reliable.
Varning: For research use only. Not for use in diagnostic procedures.
Specifikationer
  • Beskrivning:
    Agilent RNA 6000 nano kit
  • Seq:
    01
  • Details:
    For the analysis and quantitation of total and mRNA samples of 25 to 500 ng/µl in concentration, the kit includes 25 microfluidic chips, reagents and consumables, sufficient for 300 samples