CytoFLEX SRT Software Upgrades
Activation key to upgrade the CytoFLEX SRT V0-B2-Y0-R3 Cell Sorter with additional parameters. Upon installation the instrument will have 3 active lasers (488 nm, 561 nm, 638 nm) and 10 fluorescence detectors in the V0-B2-Y5-R3 configuration
Katalog # BECMC65148
Leverantör: Beckman Coulter
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:Activation key to upgrade the CytoFLEX SRT V0-B2-Y0-R3 Cell Sorter with additional parameters. Upon installation the instrument will have 3 active lasers (488 nm, 561 nm, 638 nm) and 10 fluorescence detectors in the V0-B2-Y5-R3 configuration
- Seq:12