Micropump Gear Pump Accessory Service Kit for 74445-05; 1/Ea
Catalog # MFLX74445-91
Supplier: Avantor Fluid Handling
About this item
Extend the life of your gear pump head with these service kits.
- Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
Service kit includes: gears, bushings, and seals.
- Beskrivning:Gear Pump Service Kit, for 74445-05
- Environmentally Preferable:
- Garanti:1 year
- Förp.:1