Eldex PMP PSTN 1/8" SS PM KIT
Katalog # MFLX73150-91
Leverantör: Avantor Fluid Handling
Om denna produkt
Designed for optimal operation in your Ismatec pumps.
- Choose from a wide range of replacement parts and accessories
The overall length of Solva 95605-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
- Beskrivning:1/8" Stainless Steel PM Kit for Piston Pump
- Environmentally Preferable:
- Garanti:1 year
- Förp.:1