70-mm Tote Wrench, Fiberglass-Reinforced Nylon
MFLX70607-99
Remove or tighten plugs on most industrial drums.
- Fiberglass-reinforced nylon wrenches are lightweight and easy to carry
- Use the nonsparking brass alloy models for opening drums containing flammables
- More corrosion resistant than steel versions
Use the round end for outside splines of bung caps. Use the long end for internal splines of bung caps.
Round end for outside splines of bung caps. Long end for internal splines of bung caps.
- Beskrivning:70-mm Tote Wrench, Fiberglass-Reinforced Nylon
- Längd:254 mm
- Environmentally Preferable:
- Förp.:1