Manual Purge Kit for Liquid/Solids In-Line Separators
Katalog # MFLX29995-60
Leverantör: Avantor Fluid Handling
Om denna produkt
Optimize performance of your Liquid-Solids Separator.
- Keep your Separators running properly with these kits
Choose from a wide range of replacement parts and accessories to ensure top performance of your Liquid-Solids Separator / Concentrator Units. Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.
Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.
- Beskrivning:Manual Purge Kit for Liquid/Solids In-Line Separators
- Environmentally Preferable:
- Garanti:1 year
- Kompatibel med:29995 series Liquid-Solid Separators/Concentrators
- Förp.:1