Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Thread Adapter, 10-32; 25/PK
Catalog # MFLX45518-84
Supplier: Avantor Fluid Handling
CE certificate
About this item
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
- Anslutningstyp:Male Luer to Thread
- Flow pattern:Straight
- Beskrivning:Fitting, Polypropylene, Straight, Male Luer Lock to Thread Adapter, 10-32; 25/PK
- Seq:08
- Environmentally Preferable:
- Modell:45518-84
- Connection type (port 1):Male luer lock
- Connection type (port 2):10-32 special tapered thread
- No. of ports:2
- Förp.:25
- Fitting material:Polypropylene
- Thread size UNF(M) (port 1):10-32