Masterflex® Fitting, 303 Stainless Steel, Tee, Hose Barb Union, 1/8" ID; 1EA
Katalog # MFLX31208-32
Leverantör: Avantor Fluid Handling
Om denna produkt
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/8" ID
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Hullingstorlek/slang int.Ø (port 1):1/8"
- Hullingstorlek/slang int.Ø (port 2):1/8"
- Anslutningstyp:Barbed Union
- Flow pattern:T
- Beskrivning:Fitting, 303 Stainless Steel, Tee, Hose Barb Union, 1/8" ID; 1EA
- Seq:11
- Environmentally Preferable:
- Modell:31208-32
- Orifice dimensions:0,03125"
- Garanti:1 year
- Förp.:1
- Barb size/tubing int.Ø (port 3):1/8"
- Fitting material:Stainless steel 303