Masterflex® I/P® Standard Pump Head, Replacement Rotor Assembly, Short Shaft, CRS Rotor
Katalog # MFLX07019-82
Leverantör: Avantor Fluid Handling
Om denna produkt
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Constructed of durable cold-rolled steel (CRS)
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:I/P
- Beskrivning:Standard Pump Head, Replacement Rotor Assembly, Short Shaft, CRS Rotor; 1/Ea
- Seq:26
- Environmentally Preferable:
- Modell:07019-82
- Open-head sensor:No
- Tubing sizes accepted:I/P 73, 82
- Garanti:2 years
- Förp.:1
- Rotor material:Cold-rolled steel (CRS)