Masterflex® Fitting, Polypropylene, Straight, Hose Barb Reducer, 1/4" ID x 3/16" ID; 10/PK
Catalog # MFLX40622-32
Supplier: Avantor Fluid Handling
CE certificate
About this item
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range 9 to 180 °F (-13 to +66 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Hullingstorlek/slang int.Ø (port 1):½"
- Hullingstorlek/slang int.Ø (port 2):³⁄₁₆"
- Anslutningstyp:Barbed Reducer
- Flow pattern:Straight
- Beskrivning:Fitting, polypropylene, straight, hose barb reducer, int.Ø ½×³⁄₁₆";10/pack
- Seq:28
- Environmentally Preferable:
- Modell:40622-32
- Sterilisation:Ethylene oxide (ETO) or gamma
- Förp.:10
- Fitting material:Polypropylene