Storage bottles, single-use, polystyrene, Sartolab®
250 ml, Sartolab® bottles, Genomskinlig
Katalog # 514-1295
Leverantör: Sartorius
Gå till:
Product Details & Documents
Single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.
- Low-profile design for easy gripping with one hand
- Ergonomically designed cap to provide a rigid grip and one-hand handling of each bottle
- The bottles with an indented bottom are stackable to save space in the refrigerator and in the freezer
- Caps (45 mm) with airtight seal to minimise the risk of contamination
- Graduation marks on the bottles ensure accuracy and easy reading
- Designated writing field for clear labeling of samples
- The materials used in the production are medical-grade for the highest quality, without any animal origin
- Sterile and endotoxin-free
Sartorius single-use polystyrene (PS) storage bottles are designed for safe, secure storage of tissue culture media and sera, buffers, and other aqueous solutions. The bottles can be used with all Sartolab® RF / BT vacuum filtration units with a 45 mm neck size.
Documents
Specifikationer
- Kapacitet:250 ml
- Färg:Genomskinlig
- Beskrivning:Sartolab® bottles
- Seq:02



