Tablet counting tray
14 mm, 160 mm
Catalog # SAMPA356-160
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Makes easy work of counting round tablets.
- Easy to use counting
- Suitable for all sizes of round tablets
- useful for both right-handed and left-handed operators
- Satin finish
Counting round tablets has never been easier with the Sampling Systems tablet counting triangle. When you have finished counting, the intuitive desing means the tablets can be poured directly into the bottle.
Documents
SAMPA356-275_TDS_EN
Datasheet
Specifications
- Höjd:14 mm
- Längd:160 mm
- Seq:01