Celite® 512, filterstöd, GPR RECTAPUR®
HDPE bucket, 5 kg
Leverantör: VWR Chemicals
Celite® 512, filterstöd, GPR RECTAPUR®
Katalog # 22555.360
Leverantör: VWR Chemicals
CAS-nummer: 91053-39-3
Gå till:
Product Details & Documents
Calcined Diatomaceous Earth Filter Aid.
Celite® is used as a filtering aid or as adsorbent in column chromatography.
- CAS-nummer:91053-39-3
- Formel:SiO₂
- Molecular Weight:60.08 g/mol
Documents
Certificates
Sök efter LOT-nummerP20_VWRC22555.290_SPEC_EN.pdf spec_VWRC22555.290 spec_VWRC22555.360 spec_VWRC22555.460
Produktspecifikation
VWR® for Drying and Filtration
Produktlitteratur
Specifikationer
Specifikationstestresultat
- pH (25 °C; 10%):8,5 - 11,0
- Wet density:≤ 370 g/l
- Loss on ignition (1000 °C):≤ 0,4%
- Particle size (< 75µm):≥ 90%
Specifikationer
- Förpackningstyp:HDPE bucket
- Förp.:5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



