Order Entry
Sweden
Orders LinkContactUsLinkComponent
Celite® 545, filterstöd, GPR RECTAPUR®

Celite® 545, filterstöd, GPR RECTAPUR®

PP bucket, 1 kg
Katalog # 22557.290
Irriterande
Leverantör:   VWR Chemicals
Celite® 545, filterstöd, GPR RECTAPUR®
Katalog # 22557.290
Leverantör:  VWR Chemicals
CAS-nummer:  68855-54-9
Celite® 545, filterstöd, GPR RECTAPUR®
Celite® 545, filterstöd, GPR RECTAPUR®

PP bucket, 1 kg

CAS-nummer:  68855-54-9
Price
865,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 kg
Product Details & Documents

Diatomaceous silica for filtration.

Celite® is used as a filtering aid or as adsorbent in column chromatography.

Synonymer:  DE, Diatomaceous earth, flux-calcined
  • CAS-nummer:68855-54-9
  • Formel:SiO₂
  • MDL-nummer:MFCD00132803
  • Molecular Weight:60.08 g/mol
Specifikationer

Specifikationstestresultat

  • Identification:
    Passes test
  • pH (25°C; 10%):
    ≥ 8,5
  • Loss on ignition (600 °C):
    ≤ 0.6%
  • Particle size (< 0.1 mm):
    ≥ 60%

Specifikationer

  • Förpackningstyp:
    PP bucket
  • Förp.:
    1 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.