Celite® 545, filterstöd, GPR RECTAPUR®
LDPE bag, 25 kg
Leverantör: VWR Chemicals
Celite® 545, filterstöd, GPR RECTAPUR®
Katalog # 22557.460
Leverantör: VWR Chemicals
CAS-nummer: 68855-54-9
Gå till:
Product Details & Documents
Diatomaceous silica for filtration.
Celite® is used as a filtering aid or as adsorbent in column chromatography.
Synonymer: DE, Diatomaceous earth, flux-calcined
- CAS-nummer:68855-54-9
- Formel:SiO₂
- MDL-nummer:MFCD00132803
- Molecular Weight:60.08 g/mol
Specifikationer
Specifikationstestresultat
- Identification:Passes test
- pH (25°C; 10%):≥ 8,5
- Loss on ignition (600 °C):≤ 0.6%
- Particle size (< 0.1 mm):≥ 60%
Specifikationer
- Förpackningstyp:LDPE bag
- Förp.:25 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



