Orders LinkContactUsLinkComponent
Label maker, b.safe
Label maker, b.safe
Label cassette black for b.safe label maker
Katalog # 590-0218
Leverantör:   Bohlender
Label maker, b.safe
Katalog # 590-0218
Leverantör:  Bohlender
Label maker, b.safe
Label maker, b.safe
Label cassette black for b.safe label maker
Price
740,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Portable label printer with computer-style keyboard, large LCD display and 20 methods to format text.
  • Portable
  • Powered batteries or optional AC-adapter
Powered by 6 batteries AAA or optional AC-adapter (both not included).
Leveransinformation: Includes one label cassette black on white, width 12 mm/length 3 m. Labels up to a width of 12 mm can be printed.
Specifikationer
  • Beskrivning:
    Label cassette black for b.safe label maker
  • Seq:
    02