Quick couplings, b.safe
Coupling piece, 1× UNF ¹/₄" (female) with coupling connection male, 1× fitting, Tubing Ext.Ø: 3,2 mm
Katalog # 590-0211
Leverantör: Bohlender
Gå till:
Produktdetaljer och dokument
Two-part tubing coupling made of PP with PFA Fitting to connect capillaries with Ext.Ø. 3,2 mm. The passage with the integrated quick lock is tightly closed through loosening the coupling. Tubing connections are easily interrupted without dripping from the tube, tubes and removal flasks are tightly closed during exchange. Version and scope of delivery as per below chart.
- Tightly closed
- The valves automatically open and a leakage free connection is now achieved
- If the parts are separated, the valves are automatically closed in order to avoid solvent escape
The capillary coupling is stacked together simply by hand. The coupling is successfully snapped if a clicking noise is audible. Depending on the combination of the coupling parts, the coupling can also be screwed as a capillary connection onto b.safe caps or just be used as quick connector of individual tubes.
Specifikationer
- För:Tubing Ext.Ø: 3,2 mm
- Beskrivning:Coupling piece, 1× UNF ¹/₄" (female) with coupling connection male, 1× fitting
- Seq:04