Multiports, b.safe
Tubing connector, 2× capillary connector, 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1 hose connector for tubing Int.Ø 5,5 - 8,0 mm, NPT ¹/₈"
Katalog # 590-0188
Leverantör: Bohlender
Gå till:
Produktdetaljer och dokument
Extension of the number of capillary and/or tubing connectors on a b.safe Waste Cap. With PFA Fittings for capillaries with Ext.Ø 1,6/2,2/3,2 mm (thread UNF ¹/₄"), 1 hose connector for tubing Int.Ø 5,5 to 8,0 mm (thread NPT ¹/₈").
- Excellent chemical resistance
- Available for a wide range of hose diameters
- The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary and tubing connectors of a b.safe waste cap.
Specifikationer
- Gänga:NPT ¹/₈"
- Med:2× capillary connector, 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1 hose connector for tubing Int.Ø 5,5 - 8,0 mm
- Beskrivning:Tubing connector
- Seq:02