Forceps, straight
200 mm
Katalog # USBE3035
Leverantör: USBECK Laborgeräte
Gå till:
Product Details & Documents
Made from nickel plated steel or from stainless steel.
- Straight
- Fine points
Specifikationer
- Längd:200 mm