Multiports, b.safe
Tubing connector, 3× hose connectors for tubing Int.Ø 5,5 - 8,0 mm, NPT ¹/₈"
Catalog # BOHLM509-34
Supplier: Bohlender
Jump to:
Product Details & Documents
Extension of the number of capillary and/or tubing connectors on a b.safe Waste Cap. With PFA Fittings for capillaries with Ext.Ø 1,6/2,2/3,2 mm (thread UNF ¹/₄"), 1 hose connector for tubing Int.Ø 5,5 to 8,0 mm (thread NPT ¹/₈").
- Excellent chemical resistance
- Available for a wide range of hose diameters
- The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary and tubing connectors of a b.safe waste cap.
Specifications
- Gänga:NPT ¹/₈"
- Med:3× hose connectors for tubing Int.Ø 5,5 - 8,0 mm
- Beskrivning:Tubing connector
- Seq:05
- Cat. No.:BOHLM509-34