Fluorite Objectives for Revolve Microscope
Plan fluorite water dipping; 0,5 NA, 20×, 3,5 mm
Katalog # ECHLJMAG0218
Leverantör: DISCOVER ECHO
Gå till:
Product Details & Documents
Documents
ECHO Meet Revolve
Produktlitteratur
Specifikationer
- Förstoring:20×
- Beskrivning:Plan fluorite water dipping; 0,5 NA
- Seq:22
- Arbetsavstånd:3,5 mm