ContactUsLinkComponent
ends cut Outside= 2 ± 0,1 x 1 ± 0,1 Inside= 1,78 ± 0,1 x 0,78 ± 0,1 mm L= 80 ± 0,5 wall thickness = 0,11 ± 0,05 mm in cardboard box 1 * 50 Items
Catalog # HILG1470640
Supplier:  HILGENBERG
ends cut Outside= 2 ± 0,1 x 1 ± 0,1 Inside= 1,78 ± 0,1 x 0,78 ± 0,1 mm L= 80 ± 0,5 wall thickness = 0,11 ± 0,05 mm in cardboard box 1 * 50 Items
Catalog # HILG1470640
Supplier:  HILGENBERG