ContactUsLinkComponent
TubeFiller TF3000 fast-bottle dosing tube filling unit for large tube & bottle filling, with free usable worksurface, no rack obligation. Complete set including software, 1 tubing set with filling nozzle, USB PC-connection cable 1.8 m. Ready to use / 1 * 1 Items
Catalog # BTOL970010
Leverantör:  BioTool
TubeFiller TF3000 fast-bottle dosing tube filling unit for large tube & bottle filling, with free usable worksurface, no rack obligation. Complete set including software, 1 tubing set with filling nozzle, USB PC-connection cable 1.8 m. Ready to use / 1 * 1 Items
Catalog # BTOL970010
Leverantör:  BioTool