Orders LinkContactUsLinkComponent
Objectives
Objectives
HI PLAN achromatic objective, 40×/0,65, 0,35 mm
Catalog # LEIM11506238
Supplier:   LEICA MICROSYSTEMS
Objectives
Catalog # LEIM11506238
Supplier:  LEICA MICROSYSTEMS
Objectives
HI PLAN achromatic objective, 40×/0,65, 0,35 mm
Price
4 150,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Plan achromatic and phase contrast objectives.
Specifications
  • Förstoring:
    40×/0,65
  • Beskrivning:
    HI PLAN achromatic objective
  • Arbetsavstånd:
    0,35 mm