Objectives
HI PLAN achromatic objective, 4×/0,10, 18 mm
Katalog # LEIM11506226
Leverantör: LEICA MICROSYSTEMS
Gå till:
Product Details & Documents
Plan achromatic and phase contrast objectives.
Specifikationer
- Förstoring:4×/0,10
- Beskrivning:HI PLAN achromatic objective
- Arbetsavstånd:18 mm