Surface cleaner, VHB™
VHB™ surface cleaner, 25 ml, 9×100×25 ml, Sachet
Katalog # MMMAVHB-25ML
Leverantör: 3M
Gå till:
Produktdetaljer och dokument
VHB™ surface cleaner is a very pure solution of isopropyl alcohol in water. It is colourless, has very little odour, and evaporates very quickly leaving no residue.
- Degreasing of surfaces
- Preparation of surfaces for bonding
- Evaporates quickly leaving no residue
- Recommended for use with 3M™ VHB™ Tapes
- Use to create a 'slip plane' effect to allow contact without wear
Apply this cleaner for an odourless, colourless cleaner that evaporates quickly. It leaves no residue when cleaning surfaces. It will remove most light contamination (grease, dirt) efficiently without affecting the substrate.
It cleans and degreases to ensure optimum adhesion for 3M™ VHB™ tapes due to its very pure solution of Isopropyl alcohol in water. Sold in multiple sizes, it’s also a recommended alternative to 3M™ VHB™ surface cleaner sachets.
Specifikationer
- Kapacitet:25 ml
- Beskrivning:VHB™ surface cleaner
- Seq:01
- Förpackningstyp:Sachet
- Förp.:9×100×25 ml