ContactUsLinkComponent
[EN]ROTOR FOR RVC 2-33. 48 x 15 ml 1 * 1 Items
Katalog # CHRS124538
Leverantör:  CHRIST MARTIN
[EN]ROTOR FOR RVC 2-33. 48 x 15 ml 1 * 1 Items
Katalog # CHRS124538
Leverantör:  CHRIST MARTIN