ContactUsLinkComponent
Human CD24, Mouse IgG2a Fc Tag (CD4-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P2 5063-1). 1 * 100 µG
Katalog # ACRBCD4-H5257100UG
Leverantör:  ACROBIOSYSTEMS
Human CD24, Mouse IgG2a Fc Tag (CD4-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P2 5063-1). 1 * 100 µG
Katalog # ACRBCD4-H5257100UG
Leverantör:  ACROBIOSYSTEMS