ContactUsLinkComponent
Biotinylated Human CD24, His,Avitag (CD4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accessio n P25063-1). 1 * 200 µG
Katalog # ACRBCD4-H82E9200UG
Leverantör:  ACROBIOSYSTEMS
Biotinylated Human CD24, His,Avitag (CD4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accessio n P25063-1). 1 * 200 µG
Katalog # ACRBCD4-H82E9200UG
Leverantör:  ACROBIOSYSTEMS