ContactUsLinkComponent
Recombinat Protein,Human CNTF R alpha Protein, His Tag,Human CNTF R alpha, His Tag (CNR-H5225) is expressed from human 293 cells (HE K293). It contains AA Gln 23 - Pro 346 (Accession AAH01492).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg
Katalog # ACRBCNR-H5225-1MG
Leverantör:  ACROBIOSYSTEMS
Recombinat Protein,Human CNTF R alpha Protein, His Tag,Human CNTF R alpha, His Tag (CNR-H5225) is expressed from human 293 cells (HE K293). It contains AA Gln 23 - Pro 346 (Accession AAH01492).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg
Katalog # ACRBCNR-H5225-1MG
Leverantör:  ACROBIOSYSTEMS